accessibility menu, dialog, popup

UserName

CAS RN 27511-79-1

H2NONHNH2NH2NONHNH2NOHSOHOO

CAS RN 27511-79-1

IUPAC Name: bis(5-amino-1H-pyrazole-4-carboxamide); sulfuric acid
Synonyms: bis(3-amino-2H-pyrazole-4-carboxamide); sulfuric acid
Molecular Weight (g/mol): 350.31
Molecular Formula: C8H14N8O6S
InChi Key: UMPKASYMNORSRO-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC(=O)C1=C(N)NN=C1.NC(=O)C1=C(N)NN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results