missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazole-4-carboxamide Hemisulfate 98.0+%, TCI America™

Catalog No. p-7117500
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15325G 5 g
A153225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15325G Supplier TCI America Supplier No. A15325G
Only null left

Chemical Identifiers

CAS 27511-79-1
Molecular Formula C8H14N8O6S
Molecular Weight (g/mol) 350.31
MDL Number MFCD00013093
InChI Key UMPKASYMNORSRO-UHFFFAOYSA-N
Synonym 3-amino-4-pyrazolecarboxamide hemisulfate, 5-aminopyrazole-4-carboxamide hemisulphate, 3-amino-1h-pyrazole-4-carboxamide hemisulfate, 1h-pyrazole-4-carboxamide, 3-amino-, sulfate 2:1, 3-amino-4-pyrazolecarboxamide hemisulfate salt, 3-aminopyrazole-4-carboxamide hemisulfate, 5-amino-1h-pyrazole-4-carboxamide; sulfuric acid, bis 3-amino-1h-pyrazole-4-carboxamide ; sulfuric acid, bis 3-amino-2h-pyrazole-4-carboxamide ; sulfuric acid, 5-amino-1h-pyrazole-4-carboxamide hemisulphate
PubChem CID 2723907
IUPAC Name 5-amino-1H-pyrazole-4-carboxamide;sulfuric acid
SMILES C1=NNC(=C1C(=O)N)N.C1=NNC(=C1C(=O)N)N.OS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 175.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyrazole-4-carboxamide Hemisulfate