accessibility menu, dialog, popup

UserName

CH3OOClBr

CAS RN 27007-53-0

IUPAC Name: methyl 2-bromo-5-chlorobenzoate
Synonyms: methyl 2-bromo-5-chlorobenzoate
Molecular Weight (g/mol): 249.49
Molecular Formula: C8H6BrClO2
InChi Key: BIECSXCXIXHDBC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)