accessibility menu, dialog, popup

UserName

CH3H3CCH3HN(S)HOONSNNOOHOOHO(Z)

CAS RN 26921-17-5

IUPAC Name: (2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
Synonyms: timoptic timolol hydrogen maleate (S)-timolol hydrogen maleate blocadren (S)-timolol maleate
Molecular Weight (g/mol): 432.49
Molecular Formula: C17H28N4O7S
InChi Key: WLRMANUAADYWEA-NWASOUNVSA-N
SMILES: OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (2)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results