accessibility menu, dialog, popup

UserName

CAS RN 267654-00-2

IUPAC Name: 2-phenyl-N-[5-(propan-2-yl)-1,3-thiazol-2-yl]acetamide
Synonyms: N-(5-isopropyl-1,3-thiazol-2-yl)-2-phenylacetamide
Molecular Weight (g/mol): 260.36
Molecular Formula: C14H16N2OS
InChi Key: IGRZXNLKVUEFDM-UHFFFAOYSA-N
SMILES: CC(C)C1=CN=C(NC(=O)CC2=CC=CC=C2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results