accessibility menu, dialog, popup

UserName

H3COOOOH3C(R)O(R)OH(S)OOH3C(Z)(Z)(Z)

CAS RN 26266-58-0

IUPAC Name: 2-[(2R,3S,4R)-4-hydroxy-3-[(9Z)-octadec-9-enoyloxy]oxolan-2-yl]-2-[(9Z)-octadec-9-enoyloxy]ethyl (9Z)-octadec-9-enoate
Synonyms: sorbitan trioleate
Molecular Weight (g/mol): 957.52
Molecular Formula: C60H108O8
InChi Key: PRXRUNOAOLTIEF-XDTJCZEISA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCC(OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@@H](O)[C@@H]1OC(=O)CCCCCCC\C=C/CCCCCCCC
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)