accessibility menu, dialog, popup

UserName

CAS RN 261951-78-4

IUPAC Name: 1-fluoro-2-methoxy-4-(trifluoromethyl)benzene
Synonyms: 1-fluoro-2-methoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: AXEAQZIYCQTHQV-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results