accessibility menu, dialog, popup

UserName

CH3H3CCH3OONH(R)OOBr

CAS RN 261380-20-5

IUPAC Name: (3R)-3-(4-bromophenyl)-3-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: (3R)-3-(4-bromophenyl)-3-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 343.2
Molecular Formula: C14H17BrNO4
InChi Key: ZAMLGGRVTAXBHI-LLVKDONJSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC([O-])=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results