accessibility menu, dialog, popup

UserName

CH3BrBrO

CAS RN 26074-52-2

IUPAC Name: 2-bromobutanoyl bromide
Synonyms: 2-bromobutanoyl bromide
Molecular Weight (g/mol): 229.9
Molecular Formula: C4H6Br2O
InChi Key: HHKDBXNYWNUHPL-UHFFFAOYNA-N
SMILES: CCC(Br)C(Br)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results