accessibility menu, dialog, popup

UserName

CAS RN 25808-30-4

IUPAC Name: hydrogen 2-(methylamino)acetonitrile chloride
Synonyms: hydrogen 2-(methylamino)acetonitrile chloride
Molecular Weight (g/mol): 106.55
Molecular Formula: C3H7ClN2
InChi Key: DFKBQHBEROHUNF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results