accessibility menu, dialog, popup

UserName

HBrNH2Br

CAS RN 2576-47-8

IUPAC Name: hydrogen 2-bromoethan-1-amine bromide
Synonyms: hydrogen 2-bromoethanamine bromide
Molecular Weight (g/mol): 204.89
Molecular Formula: C2H7Br2N
InChi Key: WJAXXWSZNSFVNG-UHFFFAOYSA-N
SMILES: [H+].[Br-].NCCBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)