accessibility menu, dialog, popup

UserName

CH3CH3NH3C

CAS RN 2571-52-0

IUPAC Name: 2,4,6-trimethylbenzonitrile
Synonyms: 2,4,6-trimethylbenzonitrile
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: SNIZBGQMWRHNDY-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C#N)C(C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)