accessibility menu, dialog, popup

UserName

CAS RN 2566-97-4

H3COOCH3(E)(E)

CAS RN 2566-97-4

IUPAC Name: methyl (9E,12E)-octadeca-9,12-dienoate
Synonyms: methyl (9E,12E)-octadeca-9,12-dienoate
Molecular Weight (g/mol): 294.48
Molecular Formula: C19H34O2
InChi Key: WTTJVINHCBCLGX-ZDVGBALWSA-N
SMILES: CCCCC\C=C\C\C=C\CCCCCCCC(=O)OC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results