missing translation for 'onlineSavingsMsg'
Learn More

Methyl linolelaidate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013754
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013754 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013754 Supplier Merck Emd Millipore Supplier No. 62155100MG
Only null left

Specifications

CAS 2566-97-4
Shipping Condition Ambient
Physical Form Neat
Linear Formula C19H34O2
MDL Number MFCD00069997
Synonym Linolelaidic acid methyl ester; Methyl trans,trans-9,12-octadecadienoate
Type Analytical Standard
Quantity 100 mg
Formula Weight 294.47
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less