accessibility menu, dialog, popup

UserName

CAS RN 25637-16-5

OBr

CAS RN 25637-16-5

IUPAC Name: 4-bromooxane
Synonyms: 4-bromooxane
Molecular Weight (g/mol): 165.03
Molecular Formula: C5H9BrO
InChi Key: IVBVKTPDEWDNRW-UHFFFAOYSA-N
SMILES: BrC1CCOCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results