accessibility menu, dialog, popup

UserName

CAS RN 2551-83-9

IUPAC Name: trimethoxy(prop-2-en-1-yl)silane
Synonyms: trimethoxy(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 162.26
Molecular Formula: C6H14O3Si
InChi Key: LFRDHGNFBLIJIY-UHFFFAOYSA-N
SMILES: CO[Si](CC=C)(OC)OC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)