accessibility menu, dialog, popup

UserName

CAS RN 25364-47-0

IUPAC Name: 1-[2,6-bis(propan-2-yl)phenyl]-1H-imidazole
Synonyms: 1-(2,6-diisopropylphenyl)imidazole
Molecular Weight (g/mol): 228.34
Molecular Formula: C15H20N2
InChi Key: OWSXDWUAPKBLOI-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N1C=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results