accessibility menu, dialog, popup

UserName

CAS RN 25332-39-2

IUPAC Name: hydrogen 2-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one chloride
Synonyms: desyrel
Molecular Weight (g/mol): 408.33
Molecular Formula: C19H23Cl2N5O
InChi Key: OHHDIOKRWWOXMT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=CC(=C1)N1CCN(CCCN2N=C3C=CC=CN3C2=O)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Grade 
  • (1)
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)