missing translation for 'onlineSavingsMsg'
Learn More

Trazodone hydrochloride, 98+%

Catalog No. AAJ6307006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6307006 5 g
AAJ6307003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6307006 Supplier Thermo Scientific Chemicals Supplier No. J6307006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A serotonin uptake inhibitor | CAS: 25332-39-2 | C19H23Cl2N5O | 408.327 g/mol

A serotonin uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25332-39-2
Molecular Formula C19H23Cl2N5O
Molecular Weight (g/mol) 408.327
MDL Number MFCD00079603
InChI Key OHHDIOKRWWOXMT-UHFFFAOYSA-N
Synonym trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
PubChem CID 62935
ChEBI CHEBI:9655
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
SMILES C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl

Specifications

Melting Point 222°C to 227°C
Color White
Quantity 5 g
Merck Index 14,9579
Solubility Information Soluble at 25mg/ml in methanol; Also soluble in 0;1M HCl or DMSO; Insoluble in water /n
Formula Weight 408.33
Physical Form Powder
Chemical Name or Material Trazodone hydrochloride
Hazard Category H302-H351
Hazard Statement GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 246-855-5
RTECSNumber XZ5660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only