missing translation for 'onlineSavingsMsg'
Learn More

Medisca Trazodone Hydrochloride USP

Catalog No. 14185657
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185657 25 g
14185249 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14185657 Supplier Medisca Supplier No. 055904

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trazodone Hydrochloride USP is a white to off-white, crystalline powder.

Trazodone Hydrochloride USP is a white to off-white, crystalline powder.

Specifications

Chemical Name or Material Trazodone Hydrochloride USP
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 25332-39-2
Type Drug Standard
Molecular Weight (g/mol) 408.3307
Quantity 25 g
Percent Purity 98 to 102 %
Grade USP