CAS RN 25321-09-9
IUPAC Name:
1,2-bis(propan-2-yl)benzene
Synonyms:
1,2-diisopropylbenzene
Molecular Weight (g/mol):
162.28
Molecular Formula:
C12H18
InChi Key:
OKIRBHVFJGXOIS-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC=C1C(C)C
1
–
1
of
1
results
1
Diisopropylbenzene (mixture of isomers) 97.0+%, TCI America™
CAS: 25321-09-9 Molecular Formula: C12H18 Molecular Weight (g/mol): 162.28 MDL Number: MFCD00054263 InChI Key: OKIRBHVFJGXOIS-UHFFFAOYSA-N Synonym: 1,2-diisopropylbenzene,diisopropylbenzene,o-diisopropylbenzene,benzene, 1,2-bis 1-methylethyl,benzene, o-diisopropyl,benzene, bis 1-methylethyl,unii-6e5460r9hg,diisopropylbenzene mixture of isomers,1,2-bis 1-methylethyl benzene,diisopropyl benzene PubChem CID: 11345 IUPAC Name: 1,2-bis(propan-2-yl)benzene SMILES: CC(C)C1=CC=CC=C1C(C)C