missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylbenzene (mixture of isomers) 97.0+%, TCI America™

Catalog No. D092925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D092925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D092925ML Supplier TCI America Supplier No. D092925ML
Only null left

Chemical Identifiers

CAS 25321-09-9
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00054263
InChI Key OKIRBHVFJGXOIS-UHFFFAOYSA-N
Synonym 1,2-diisopropylbenzene, diisopropylbenzene, o-diisopropylbenzene, benzene, 1,2-bis 1-methylethyl, benzene, o-diisopropyl, benzene, bis 1-methylethyl, unii-6e5460r9hg, diisopropylbenzene mixture of isomers, 1,2-bis 1-methylethyl benzene, diisopropyl benzene
PubChem CID 11345
IUPAC Name 1,2-bis(propan-2-yl)benzene
SMILES CC(C)C1=CC=CC=C1C(C)C

Specifications

Color Colorless
Quantity 25 mL
UN Number 3082
Formula Weight 162.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropylbenzene (mixture of isomers)
EINECSNumber (3)-0025
RTECSNumber CZ6330000
TSCA Yes