CAS RN 252938-22-0
IUPAC Name:
1,3-dimethyl 2-[(4-fluorophenyl)methyl]propanedioate
Synonyms:
1,3-dimethyl 2-[(4-fluorophenyl)methyl]propanedioate
Molecular Weight (g/mol):
240.23
Molecular Formula:
C12H13FO4
InChi Key:
WDFUURBBGZTSIA-UHFFFAOYSA-N
SMILES:
COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC
1
–
1
of
1
results
1
Promotions Available
Dimethyl (4-fluorobenzyl)malonate, 97%
Dimethyl (4-fluorobenzyl)malonate, 97%
CAS: 252938-22-0 Molecular Formula: C12H13FO4 Molecular Weight (g/mol): 240.23 MDL Number: MFCD01934558 InChI Key: WDFUURBBGZTSIA-UHFFFAOYSA-N Synonym: dimethyl 4-fluorobenzyl malonate,dimethyl 2-4-fluorobenzyl malonate,1,3-dimethyl 2-4-fluorophenyl methyl propanedioate,acmc-1cpd2,dimethyl 4-fluorobenzylmalonate,2-4-fluorobenzyl-malonic acid dimethyl ester,dimethyl 2-4-fluorophenyl methyl propanedioate,dimethyl 2-4-fluorophenyl methyl propane-1,3-dioate PubChem CID: 2781576 SMILES: COC(=O)C(CC1=CC=C(F)C=C1)C(=O)OC