accessibility menu, dialog, popup

UserName

H3COON

CAS RN 2524-52-9

IUPAC Name: ethyl pyridine-2-carboxylate
Synonyms: picolinic acid, ethyl ester
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: FQYYIPZPELSLDK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=N1
Boiling Point 
  • (4)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (4)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)