accessibility menu, dialog, popup

UserName

CAS RN 25179-23-1

IUPAC Name: lithium(1+) 2-methylpropanoate
Synonyms: lithium(1+) isobutyrate
Molecular Weight (g/mol): 94.04
Molecular Formula: C4H7LiO2
InChi Key: IIDVGIFOWJJSIJ-UHFFFAOYSA-M
SMILES: [Li+].CC(C)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results