accessibility menu, dialog, popup

UserName

CAS RN 25135-51-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C27H22O4S)n
InChi Key: —
SMILES: CC(C)(C1=CC=C(O-*)C=C1)C1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(-*)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results