accessibility menu, dialog, popup

UserName

CAS RN 2511-22-0

IUPAC Name: methyl 3,5-di-tert-butyl-4-hydroxybenzoate
Synonyms: methyl 3,5-di-tert-butyl-4-hydroxybenzoate
Molecular Weight (g/mol): 264.37
Molecular Formula: C16H24O3
InChi Key: UPVYFJALDJUSOV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results