accessibility menu, dialog, popup

UserName

Br

CAS RN 25109-28-8

IUPAC Name: 1-bromo-4-cyclohexylbenzene
Synonyms: 1-bromo-4-cyclohexylbenzene
Molecular Weight (g/mol): 239.16
Molecular Formula: C12H15Br
InChi Key: LVIJLEREXMVRAN-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)