missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-cyclohexylbenzene 97.0+%, TCI America™

Catalog No. B26865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26865G 5 g
B268625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26865G Supplier TCI America Supplier No. B26865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25109-28-8
Molecular Formula C12H15Br
Molecular Weight (g/mol) 239.156
InChI Key LVIJLEREXMVRAN-UHFFFAOYSA-N
Synonym p-bromocyclohexylbenzene, 1-brom-4-cyclohexylbenzol, zlchem 88, 4-cyclohexylbromobenzene, 4-bromophenylcyclohexane, acmc-1ccsm, 4-bromo-1-cyclohexylbenzene, 1-bromo-4-cyclohexyl-benzene, benzene, 1-bromo-4-cyclohexyl, lvijlerexmvran-uhfffaoysa
PubChem CID 90718
IUPAC Name 1-bromo-4-cyclohexylbenzene
SMILES C1CCC(CC1)C2=CC=C(C=C2)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 239.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-cyclohexylbenzene