accessibility menu, dialog, popup

UserName

H3COBrO

CAS RN 25016-01-7

IUPAC Name: 5-bromo-2-methoxybenzaldehyde
Synonyms: 5-bromo-2-methoxybenzaldehyde
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: IJIBRSFAXRFPPN-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1C=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)