missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-o-anisaldehyde 98.0+%, TCI America™

Catalog No. B179125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B179125G 25 g
B1791250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B179125G Supplier TCI America Supplier No. B179125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25016-01-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00003347
InChI Key IJIBRSFAXRFPPN-UHFFFAOYSA-N
Synonym 5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
PubChem CID 90684
IUPAC Name 5-bromo-2-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)Br)C=O

Specifications

Melting Point 117°C
Quantity 25 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-o-anisaldehyde