missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-anisaldehyde, 99%

Catalog No. AC167521000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167521000 100 g Glass bottle
AC167520050 5 g Glass bottle
AC167520250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167521000 Supplier Thermo Scientific Chemicals Supplier No. 167521000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25016-01-7 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25016-01-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00003347
InChI Key IJIBRSFAXRFPPN-UHFFFAOYSA-N
Synonym 5-bromo-2-anisaldehyde, 5-bromo-o-anisaldehyde, 2-methoxy-5-bromobenzaldehyde, benzaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxy-benzaldehyde, pubchem2644, acmc-209ggg, intermediates-zcf02672, o-anisaldehyde, 5-bromo, ksc494q2j
PubChem CID 90684
IUPAC Name 5-bromo-2-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)Br)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 118°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(OCH3)CHO
Quantity 100 g
Beilstein 08, 55
Formula Weight 215.05
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-anisaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 246-564-3

RUO – Research Use Only