CAS RN 2492-26-4
IUPAC Name:
sodium (1,3-benzothiazol-2-yl)sulfanide
Synonyms:
sodium 1,3-benzothiazol-2-ylsulfanide
Molecular Weight (g/mol):
189.23
Molecular Formula:
C7H4NNaS2
InChi Key:
VLDHWMAJBNWALQ-UHFFFAOYSA-M
SMILES:
[Na+].[S-]C1=NC2=CC=CC=C2S1
1
–
1
of
1
results
1
Sodium 2-Mercaptobenzothiazole 97.0+%, TCI America™
CAS: 2492-26-4 Molecular Formula: C7H5NNaS2 Molecular Weight (g/mol): 190.234 MDL Number: MFCD00065334 InChI Key: KRXFTOUYGXMRRU-UHFFFAOYSA-N Synonym: Sodium 2-Benzothiazolethiolate, 2-Mercaptobenzothiazole Sodium Salt PubChem CID: 24195611 IUPAC Name: 3H-1,3-benzothiazole-2-thione;sodium SMILES: C1=CC=C2C(=C1)NC(=S)S2.[Na]