CAS RN 2482-57-7
IUPAC Name:
1-bromobutan-2-ol
Synonyms:
1-bromobutan-2-ol
Molecular Weight (g/mol):
153.02
Molecular Formula:
C4H9BrO
InChi Key:
DMRXISNUOWIOKV-UHFFFAOYNA-N
SMILES:
CCC(O)CBr
1
–
1
of
1
results
1
1-Bromo-2-butanol (contains ca. 20% 2-Bromo-1-butanol) 73.0+%, TCI America™
CAS: 2482-57-7 Molecular Formula: C4H9BrO Molecular Weight (g/mol): 153.019 MDL Number: MFCD00059883 InChI Key: DMRXISNUOWIOKV-UHFFFAOYSA-N Synonym: 1,2-Butane Bromohydrin PubChem CID: 520149 IUPAC Name: 1-bromobutan-2-ol SMILES: CCC(CBr)O