accessibility menu, dialog, popup

UserName

CAS RN 247570-24-7

IUPAC Name: tert-butyl N-(4-aminocyclohexyl)carbamate
Synonyms: tert-butyl N-(4-aminocyclohexyl)carbamate
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: FEYLUKDSKVSMSZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results