accessibility menu, dialog, popup

UserName

CAS RN 2475-81-2

IUPAC Name: methyl 2-amino-4-fluorobenzoate
Synonyms: methyl 2-amino-4-fluorobenzoate
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: UBFRSTYHLYPSND-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results