accessibility menu, dialog, popup

UserName

CAS RN 2451-62-9

IUPAC Name: tris[(oxiran-2-yl)methyl]-1,3,5-triazinane-2,4,6-trione
Synonyms: tris(oxiran-2-ylmethyl)-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol): 297.27
Molecular Formula: C12H15N3O6
InChi Key: OUPZKGBUJRBPGC-UHFFFAOYNA-N
SMILES: O=C1N(CC2CO2)C(=O)N(CC2CO2)C(=O)N1CC1CO1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results