accessibility menu, dialog, popup

UserName

H2ONH2NNNN(R)O(R)OH(S)HO(S)HO

CAS RN 24356-66-9

IUPAC Name: (2R,3S,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol hydrate
Synonyms: ara-A arabinoside adenine arabinosyladenine triphosphate arabinosyladenine ara-a triphosphate arabinosyl adenine Ara-Atp arabinosyl-atp arabinofuranosyladenine triphosphate ara atp
Molecular Weight (g/mol): 285.26
Molecular Formula: C10H15N5O5
InChi Key: ZTHWFVSEMLMLKT-CAMOTBBTSA-N
SMILES: O.NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)C2=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results