accessibility menu, dialog, popup

UserName

CAS RN 24342-68-5

IUPAC Name: 1-phenyl-2,5,8,11,14,17-hexaoxanonadecan-19-ol
Synonyms: 1-phenyl-2,5,8,11,14,17-hexaoxanonadecan-19-ol
Molecular Weight (g/mol): 372.46
Molecular Formula: C19H32O7
InChi Key: VVBQKDDPSXBMMZ-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCCOCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results