missing translation for 'onlineSavingsMsg'
Learn More

Hexaethylene glycol monobenzyl ether

Catalog No. AC465840010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465840010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC465840010 Supplier Thermo Scientific Chemicals Supplier No. 465840010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24342-68-5 | C19H32O7 | 372.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24342-68-5
Molecular Formula C19H32O7
Molecular Weight (g/mol) 372.46
InChI Key VVBQKDDPSXBMMZ-UHFFFAOYSA-N
IUPAC Name 1-phenyl-2,5,8,11,14,17-hexaoxanonadecan-19-ol
SMILES OCCOCCOCCOCCOCCOCCOCC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 372.46
Chemical Name or Material Hexaethylene glycol monobenzyl ether
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only