missing translation for 'onlineSavingsMsg'
Learn More

Hexaethylene Glycol Monobenzyl Ether 93.0+%, TCI America™

Catalog No. H10431G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10431G Supplier TCI America Supplier No. H10431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24342-68-5
Molecular Formula C19H32O7
Molecular Weight (g/mol) 372.458
MDL Number MFCD06797107
InChI Key VVBQKDDPSXBMMZ-UHFFFAOYSA-N
Synonym 2-[2-[2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
PubChem CID 11164646
IUPAC Name 2-[2-[2-[2-[2-(2-phenylmethoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES C1=CC=C(C=C1)COCCOCCOCCOCCOCCOCCO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 372.46
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexaethylene Glycol Monobenzyl Ether