accessibility menu, dialog, popup

UserName

OBrF

CAS RN 242812-04-0

IUPAC Name: 1-[3-(bromomethyl)phenoxy]-2-fluorobenzene
Synonyms: 1-[3-(bromomethyl)phenoxy]-2-fluorobenzene
Molecular Weight (g/mol): 281.12
Molecular Formula: C13H10BrFO
InChi Key: FWAHWPGVFDHNCH-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1OC1=CC=CC(CBr)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results