missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Fluorophenoxy)benzyl Bromide, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 281.12 |
| Physical Form | Liquid |
| Chemical Name or Material | 3-(2-Fluorophenoxy)benzyl Bromide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |