accessibility menu, dialog, popup

UserName

CAS RN 24249-95-4

H3CCH2OONH3CCH3OO

CAS RN 24249-95-4

IUPAC Name: 3-[dimethyl({2-[(2-methylprop-2-enoyl)oxy]ethyl})azaniumyl]propanoate
Synonyms: 3-[dimethyl({2-[(2-methylprop-2-enoyl)oxy]ethyl})ammonio]propanoate
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: CSWRCKVZMMKVDC-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC[N+](C)(C)CCC([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results