accessibility menu, dialog, popup

UserName

CAS RN 24155-42-8

HONNClCl

CAS RN 24155-42-8

IUPAC Name: 1-(2,4-dichlorophenyl)-2-(1H-imidazol-1-yl)ethan-1-ol
Synonyms: 1-(2,4-dichlorophenyl)-2-(imidazol-1-yl)ethanol
Molecular Weight (g/mol): 257.11
Molecular Formula: C11H10Cl2N2O
InChi Key: UKVLTPAGJIYSGN-UHFFFAOYNA-N
SMILES: OC(CN1C=CN=C1)C1=CC=C(Cl)C=C1Cl
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)