accessibility menu, dialog, popup

UserName

CAS RN 2411-89-4

IUPAC Name: 2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
Synonyms: O-cresolphthalein complexone
Molecular Weight (g/mol): 636.61
Molecular Formula: C32H32N2O12
InChi Key: IYZPEGVSBUNMBE-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
Grade 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results