accessibility menu, dialog, popup

UserName

CAS RN 2409-52-1

H3COOCH2OOCH3

CAS RN 2409-52-1

IUPAC Name: 1,4-diethyl 2-methylidenebutanedioate
Synonyms: 1,4-diethyl 2-methylidenebutanedioate
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: ZEFVHSWKYCYFFL-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=C)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results