accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 2396-80-7

IUPAC Name: methyl hex-5-enoate
Synonyms: methyl hex-5-enoate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: ASKDFGVMJZMYEM-UHFFFAOYSA-N
SMILES: COC(=O)CCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results