accessibility menu, dialog, popup

UserName

FBrF

CAS RN 23915-07-3

IUPAC Name: 1-(bromomethyl)-2,4-difluorobenzene
Synonyms: 1-(bromomethyl)-2,4-difluorobenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: IBLMYGXJKQIGSN-UHFFFAOYSA-N
SMILES: FC1=CC=C(CBr)C(F)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)