missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzyl bromide, 98+%

Catalog No. AAA1521314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1521314 25 g
AAA1521306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1521314 Supplier Thermo Scientific Chemicals Supplier No. A1521314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23915-07-3 | C7H5BrF2 | 207.018 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23915-07-3
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD00011649
InChI Key IBLMYGXJKQIGSN-UHFFFAOYSA-N
Synonym 2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911
PubChem CID 90297
IUPAC Name 1-(bromomethyl)-2,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)F)CBr

Specifications

Density 1.613
Boiling Point 42°C to 43°C (1 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.525
Quantity 25 g
UN Number UN1760
Beilstein 4177539
Formula Weight 207.02
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorobenzyl bromide
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H317-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 245-938-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only